Learn More
Waters Corp Waters ACQUITY UPLC CSH Phenyl-Hexyl Column, 1.7 μm, 130 Å, 1.0 × 150 mm
Supplier:  Waters Corp 186005403
Encompass_Preferred
The ACQUITY UPLC CSH Phenyl-Hexyl column is designed for optimal performance in high-speed applications. It provides superior peak shape for basic analytes when used with formic acid or other low-ionic-strength, acidic mobile phases.
  • Provides superior peak shape for basic analytes when used with formic acid or other low-ionic-strength, acidic mobile phases.
  • Features a controlled, low-level, positive surface charge bonded to ethylene-bridged hybrid (BEH) particles.
  • Eliminates the need for ion-pairing reagents for good peak shape for bases.
  • Known for its ruggedness.
Catalog No. 50-791-419