Learn More
Waters Corp Waters ACQUITY UPLC CSH Phenyl-Hexyl Column, 1.7 μm, 130 Å, 1.0 x 100 mm
Supplier: Waters Corp 176002159
ACQUITY UPLC CSH Columns are designed to provide superior peak shape for basic analytes, particularly when used with formic acid or other low-ionic-strength, acidic mobile phases. This performance is achieved because the columns utilize a controlled, low-level, positive surface charge bonded to ethylene-bridged hybrid (BEH) particles, eliminating the need for ion-pairing reagents.
- Particle size: 1.7 μm
- Pore size: 130 Å
- Column dimensions: 1.0 × 100 mm
- Pack quantity: 3 columns
- Ligand density: 2.3 μmol/m²
- Carbon load: 14%
- Endcapped: Yes
- USP class no.: L11
- pH range: 1–11
- Temperature limits: Low pH = 80 °C, high pH = 45 °C
- Surface area: 185 m²/g
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.