Learn More
Waters Corp Waters CSH Phenyl-Hexyl Column, 130Å, 1.7 µm, 2.1 mm X 150 mm
Supplier:  Waters Corp 176002165
Encompass_Preferred
ACQUITY CSH Phenyl-Hexyl columns provide alternative analyte selectivity and are valuable for method development. The trifunctionally bonded C6 phenyl ligand acts as a robust, low bleed sorbent that selectively retains polyaromatic compounds through pi-pi interactions.
  • Provides alternative analyte selectivity
  • Valuable tool for method development
  • Trifunctionally bonded C6 phenyl ligand
  • Robust
  • Low bleed sorbent
  • Selectively retains polyaromatic compounds through pi-pi interactions
Catalog No. 50-791-130