Learn More
Waters Corp Waters ACQUITY UPLC HSS Cyano (CN) Column, 100Å, 1.8 μm, 1 mm X 100 mm
Supplier:  Waters Corp 186005983
Encompass_Preferred
ACQUITY UPLC HSS CN chemistry is an ultra-performance general purpose propyl cyano bonded phase that provides superior peak shape for bases, increased retention, and extremely long column lifetimes at low pH. ACQUITY HSS CN columns are fully scalable and transferable to HPLC separations.
  • Ultra-performance general purpose propyl cyano bonded phase
  • Provides superior peak shape for bases
  • Increased retention
  • Extremely long column lifetimes at low pH
  • Fully scalable and transferable to HPLC separations
Catalog No. 50-179-448