Learn More
Waters Corp ACQUITY UPLC HSS Cyano (CN) Column, 100Å, 1.8 µm, 2.1 mm X 30 mm, 1/pk
Supplier:  Waters Corp 186005985
Encompass_Preferred
ACQUITY UPLC HSS CN chemistry is an ultra-performance general purpose propyl cyano bonded phase that provides superior peak shape for bases, increased retention, and extremely long column lifetimes at low pH. ACQUITY HSS CN columns are fully scalable and transferable to HPLC separations.
Catalog No. 50-179-450