Learn More
Waters Corp Waters Column, Cyano (CN), 100 Å, 1.8 μm, 3 mm X 50 mm
Supplier:  Waters Corp 186005991
Encompass_Preferred
This column chemistry is an ultra-performance general purpose propyl cyano bonded phase that provides superior peak shape for bases, increased retention, and extremely long column lifetimes at low pH. These columns are fully scalable and transferable to HPLC separations.
  • Superior peak shape for bases.
  • Increased retention.
  • Extremely long column lifetimes at low pH.
  • Fully scalable and transferable to HPLC separations.
  • Made in a cGMP, ISO 9001:2000 certified plant.
  • Low hydrophobicity and unique selectivity.
  • Ultra-stable retention, exceptional peak shape, and reproducibility under low-to mid-pH conditions.
  • Compatible with both reversed-phase and normal-phase techniques.
  • Operating pH range of 2-8.
Catalog No. 50-179-455