Learn More
Waters Corp Waters ACQUITY Premier Protein SEC Column, 250Å, 1.7 μm, 4.6 x 150 mm
Supplier:  Waters Corp 176005071
Encompass_Preferred
ACQUITY Premier Protein SEC 250 Å, 1.7 μm Columns provide reliable protein size variant separations (10,000 to approximately 650,000 Daltons). They incorporate MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies to minimize secondary ionic or hydrophobic interactions. This reduces method development time, enabling a “generic” or “platform-type” method for various samples. Designed for UPLC SEC applications, requiring a low LC system volume for optimal performance.
  • Achieves reliable protein size variant separations.
  • Minimizes secondary ionic or hydrophobic interactions using MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies.
  • Reduces costly method development by enabling a “generic” or “platform-type” method.
  • Includes a standard for size variant analysis.
Catalog No. NC2067194