Learn More
Waters Corp Waters Column, BEH Amide, 1.7 μm, 2.1 mm x 50 mm
Supplier:  Waters Corp 186009504
Encompass_Preferred
These columns retain extremely polar compounds, including carbohydrates and sugars. Based on BEH technology, they feature a chemically stable, trifunctionally bonded amide phase with stability across a pH range of 2 to 11. This allows separation of polar analytes with diverse polarity, structural characteristics, and pKa values. They also incorporate MaxPeak High Performance Surface technology, reducing unwanted analyte/surface interactions that cause poor peak shape and diminished signal intensity.
  • Retains extremely polar compounds like carbohydrates and sugars.
  • Chemically stable, trifunctionally bonded amide phase.
  • Stable from pH 2 to 11 for broad polar analyte separation.
  • MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions.
  • Reduces poor peak shape and signal intensity loss.
Catalog No. 50-222-4220