Learn More
Waters Corp ACQUITY Column, Premier BEH C18, 1.7 μm, 2.1 mm, 50 mm
Supplier:  Waters Corp 186009455
Encompass_Preferred
This Premier BEH C18 Column offers high-performance separations and is suitable for method development. Its MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions, preventing poor peak shape and signal intensity losses. The trifunctionally bonded BEH particle provides a wide usable pH range (pH 1 to 12) and increased stability with low-pH mobile phases.
  • High performance separations for method development.
  • MaxPeak High Performance Surface technology reduces unwanted analyte/surface interactions.
  • Trifunctionally bonded BEH particle offers wide usable pH range (pH 1 to 12).
  • VanGuard FIT design protects column from contaminants, extending lifetime.
  • Robust and adaptable for peptide separations in UPLC systems.
  • BEH particle ensures high stability and controls silanol activity, enhancing data reproducibility.
Catalog No. 50-203-9062