Learn More
Waters Corp ACQUITY Premier HSS PFP Column, 1.8 μm, 2.1 x 100 mm, 100 Å
Supplier:  Waters Corp 186010037
Encompass_Preferred
This column features MaxPeak High Performance Surfaces (HPS) to enhance chromatographic separations by reducing analyte-surface interactions. This technology improves analyte recovery, sensitivity, and reproducibility. Its high strength silica (HSS) particle technology delivers increased resolution, speed, and sensitivity for analytical challenges.
  • MaxPeak high performance surfaces minimize analyte/surface interactions.
  • Mitigates loss of metal-sensitive analytes.
  • HSS technology offers increased retentivity and wide selectivity space.
  • High strength silica provides mechanical stability.
  • Reduces column conditioning and passivation times.
  • Improves sensitivity and peak shapes.
  • Improves recovery.
  • Allows simpler mobile phases, eliminating complex additives.
  • Increases retention and efficiency for analytes.
  • Enables diverse chromatographic interactions.
Catalog No. 50-239-1558