Learn More
Waters Corp ACQUITY Premier Peptide BEH C18 Column, 130 Å 1.7 μm, 2.1 x 100 mm
Supplier:  Waters Corp 186010697
Encompass_Preferred
The ACQUITY Premier Peptide BEH C18 Column with MaxPeak High Performance Surfaces is engineered to optimize chromatographic separations. It achieves this by minimizing analyte-surface interactions, which in turn enhances analyte recovery, sensitivity, and reproducibility.
  • Reduced column conditioning and passivation times.
  • Improved sensitivity and peak shapes.
  • Supports simpler mobile phases.
  • Universal column choice for a wide variety of compounds.
  • Exceptional peak shape for basic analytes at elevated pH.
  • Stable across a wide pH range and at high temperatures.
  • Outstanding peak capacity and superior peak shape for peptide analysis in TFA, DFA, and FA.
  • Outstanding peak capacity, superior peak shape, and lifetime for oligonucleotide analysis in HFIP, HAA, and TEA.
Catalog No. 50-239-1152