Learn More
Waters Corp C18 Column, 130 Å, 1.7 μm, 2.1 × 150 mm
Supplier:  Waters Corp 186010699
Encompass_Preferred
This column is optimized for peptide separations with a small-pore (130Å) trifunctionally bonded BEH particle. It offers a wide usable pH range, superior low pH stability, and ultra-low column bleed, suitable for proteomics and peptide synthesis. As part of the MaxPeak Premier class, it provides consistent performance, enhanced reproducibility, peak shape, and recovery for metal-sensitive analytes. An integrated, easily replaceable 5 mm pre-column protects the analytical column.
  • Optimized and QC tested for peptide separations.
  • Wide pH range and superior low pH stability (130Å BEH particle).
  • Ultra-low column bleed.
  • Enhanced reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Integrated, replaceable 5 mm pre-column for column protection.
  • Easily replaceable VanGuard FIT cartridges with qualified sorbent.
Catalog No. 50-239-2818