Learn More
Waters Corp ACQUITY Column Kit, Peptide BEH C18, 130Å, 1.7 μm, 2.1 × 150 mm
Supplier:  Waters Corp 186009659
Encompass_Preferred
The ACQUITY Premier Peptide BEH C18, 130Å, 1.7 μm 2.1 × 150 mm Columns are optimized for peptide separations. They feature a small-pore (130Å) trifunctionally bonded BEH particle, offering wide pH range, superior low pH stability, and ultra-low column bleed. Ideal for UPLC/UHPLC and LC-MS methods, enhancing reproducibility and recovery. Method validation kit (MVK) includes three columns.
  • Optimized for peptide separations.
  • Small-pore (130Å) BEH particle.
  • Wide pH range, low pH stability, ultra-low column bleed.
  • UPLC/UHPLC and LC-MS compatible.
  • Enhances reproducibility and recovery for metal-sensitive analytes.
  • MVK: three columns included.
Catalog No. 50-203-9143
Add to Cart
Request a Quote