Learn More
Waters Corp ACQUITY Premier Peptide CSH C18 Column Kit, 130Å, 1.7 μm, 2.1 x 100 mm
Supplier:  Waters Corp 186009652
Encompass_Preferred
This column is designed for peptide separations. Its 130Å, 1.7 μm, 2.1 x 100 mm MVK particles ensure excellent peak shape for peptides using FA- or TFA-containing mobile phases. Suitable for LC or LC-MS, this premier class column enhances reproducibility and recovery for metal-sensitive analytes. The method validation kit (MVK) includes three columns.
  • For peptide separations.
  • Excellent peak shape with FA- or TFA-containing mobile phases.
  • Low dependence on strong ion pairing agents (LC or LC-MS).
  • Improved reproducibility and recovery for metal-sensitive analytes.
  • MVK includes three columns of different lots.
  • QC tested with cytochrome c digest standards.
  • Ideal for low dispersion UPLC/UHPLC (2.1 mm ID).
Catalog No. 50-239-1159
Add to Cart
Request a Quote