Learn More
Waters Corp ACQUITY Premier Peptide CSH C18 Column Kit, 130Å, 1.7 μm, 2.1 x 150 mm, 3 pk
Supplier: Waters Corp 186009653
This column is designed for peptide separations, offering excellent peak shape and high reproducibility for metal-sensitive analytes. It is suitable for LC or LC-MS applications on UPLC or UHPLC systems. This Method Validation Kit includes three columns.
- Designed for peptide separations.
- Achieves excellent peak shape with FA- or TFA-containing mobile phases.
- Suitable for LC or LC-MS applications.
- Recommended for low dispersion UPLC or UHPLC systems.
- Enhances reproducibility, peak shape, and recovery for metal-sensitive analytes.
- Method Validation Kit includes three columns, each with a different material lot.
- C18 chemistry.
- Particle size: 1.7 μm.
- Pore size: 130 Å.
- Inner diameter: 2.1 mm.
- Length: 150 mm.
- Maximum pressure: 18000 psi (1240 Bar).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.