Learn More
Waters Corp Column, Peptide CSH C18, 1.7 μm, 2.1 x 50 mm
Supplier:  Waters Corp 186010709
Encompass_Preferred
This column is optimized for peptide separations, delivering excellent peak shape for peptides using FA- or TFA-containing mobile phases. It is suitable for LC or LC-MS applications and enhances reproducibility, peak shape, and recovery for metal-sensitive analytes. An integrated, replaceable 5 mm pre-column protects the analytical column.
  • Optimized for peptide separations, analysis, and mapping.
  • Provides excellent peak shape for peptides with FA- or TFA-containing mobile phases.
  • Enhances reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Features a fully integrated and easily replaceable 5 mm pre-column to protect the analytical column.
Catalog No. 50-239-2497