Learn More
Waters Corp Waters ACQUITY Premier Oligonucleotide C18 Column, 130Å, 1.7 μm, 2.1 × 50 mm
Supplier:  Waters Corp 186009484
Encompass_Preferred
ACQUITY Premier Oligonucleotide Columns are designed for separating detritylated synthetic oligonucleotides via ion-pair, reversed-phase chromatography. These MaxPeak Premier Class columns enhance reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Reduces column conditioning and passivation, simplifying mobile phases.
  • Features tri-functionally bonded BEH particle technology for accelerated method development.
  • Delivers excellent peak shape for basic analytes at elevated pH, stable across a pH range of 1–12 and high temperatures.
  • Manufactured with ultrapure reagents, ensuring controlled chemical composition and purity.
  • QC tested with MassPrep OST standards for repeatable performance; Certificates of Batch Analysis and Column Efficiency included.
  • Key specifications: 1.7 μm particle size, 130 Å pore size, 18000 psi (1240 Bar) max pressure.
Catalog No. NC1834726