Learn More
Waters Corp Waters Corporation Column, C18, 1.7 μm, 2.1 mm, 50 mm
Supplier:  Waters Corp 186009487
Encompass_Preferred
This ACQUITY Premier Peptide CSH C18 Column is designed for peptide separations, featuring 130Å, 1.7 μm particles with low, carefully defined positive charges. It provides excellent peak shape for peptides with FA- or TFA-containing mobile phases. Minimally dependent on strong ion pairing agents, it suits LC or LC-MS applications. As part of the MaxPeak Premier Class, it enhances reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Optimized for peptide separations and analysis.
  • Ensures excellent peak shape for peptides.
  • Ideal for LC or LC-MS applications due to minimal ion pairing agent dependence.
  • Recommended for low dispersion (UPLC/UHPLC) systems, especially for 2.1 mm ID columns.
  • Improves reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Quality controlled with cytochrome c digest standards.
Catalog No. NC1837186