Learn More
Waters Corp Waters Corporation ACQUITY UPC2 BEH Column, 2.1 × 100 mm, 1.7 μm, 130 Å
Supplier:  Waters Corp 186006560
Encompass_Preferred
This ACQUITY UPC2 BEH column is engineered for effective chromatographic separations, suitable for both chiral and achiral applications. It offers precise dimensions and an optimized particle size to deliver high-resolution results.
  • Designed for chiral and achiral separations.
  • Features dimensions of 2.1 × 100 mm.
  • Equipped with a particle size of 1.7 μm.
  • Utilizes BEH (Ethylene Bridged Hybrid) technology.
  • Pore size of 130 Å.
Catalog No. 50-466-316