Learn More
Waters Corp Viridis BEH Column, 130Å, 3.5 µm, 3 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186006641
Encompass_Preferred
The Viridis sub-2-µm and 3.5 µm columns demonstrate high efficiency and resolution. The reduction and control of surface silanol activity on Viridis particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.
Catalog No. 50-466-392