Learn More
Waters Corp Waters Corporation Column, CSH Fluoro-Phenyl, 3.0 x 50 mm, 3.5 μm
Supplier:  Waters Corp 186006647
Encompass_Preferred
The ACQUITY UPC2 CSH Fluoro-Phenyl column is designed for use with the ACQUITY UPC2 System, employing compressed liquid CO2 as the primary mobile phase for Convergence Chromatography. This column, with dimensions of 3.0 x 50 mm and a particle size of 3.5 μm, is intended for separating a wide range of compounds and is compatible with mass spectrometry detection.
  • Designed for use with the ACQUITY UPC2 system, utilizing compressed liquid CO2 as the primary mobile phase for convergence chromatography.
  • Intended for separating a wide range of compounds.
  • Compatible with mass spectrometry detection.
Catalog No. 50-466-398