Learn More
Waters Corp Viridis HSS C18 SB Column, 100Å, 1.8 μm, 2.1 mm x 50 mm
Supplier:  Waters Corp 186006617
Encompass_Preferred
This HSS C18 SB Column is designed for ACQUITY UPC2, providing high efficiency and resolution. It offers excellent peak shapes under SuperCritical Fluid (SFC) conditions for basic achiral compounds without mobile-phase additives, by controlling surface silanol activity. It is highly reproducible, predictable, and suitable for preparative analytical and purification.
  • Efficiency and resolution with sub-2-μm and 3.5 μm particles.
  • Good peak shapes for basic achiral compounds without mobile-phase additives under SFC.
  • Reproducible and predictable due to stringent specifications, low metal content, and silica production knowledge.
  • Usable for preparative analytical and purification columns.
  • Exhibits achiral SFC selectivities.
  • Utilizes unique Ethylene Bridged Hybrid (BEH), Charged Surface Hybrid (CSH), and High-Strength Silica (HSS) particle technologies.
Catalog No. 50-466-368