Learn More
Waters Corp Waters ACQUITY UPLC Column, BEH Amide, 1.7 μm, 1 mm X 50 mm
Supplier:  Waters Corp 176001914
Encompass_Preferred
ACQUITY BEH Amide columns are designed for Hydrophilic Interaction Chromatography (HILIC) to retain and separate extremely polar compounds, such as carbohydrates and sugars. Utilizing BEH technology, they offer a chemically stable, trifunctionally bonded amide phase for effective separation of polar analytes across a wide pH range.
  • Retains extremely polar compounds like carbohydrates and sugars.
  • Chemically stable from pH 2 to 11 and at high temperatures.
  • Features MaxPeak High-Performance Surface technology.
  • Provides excellent peak shapes for bases without ion-pairing reagents.
  • Enables high-resolution, high-speed analysis of polar analytes.
  • Increases mass spectrometry response and simplifies detection.
Catalog No. 50-465-908