Learn More
Waters Corp ACQUITY UPLC BEH Amide Column, 130Å, 1.7 µm, 1 mm X 50 mm, 1/pk
Supplier:  Waters Corp 186004848
Encompass_Preferred
ACQUITY BEH Amide columns retain extremely polar compounds, including carbohydrates and sugars. Based on BEH technology the ACQUITY Amide columns offer a chemically stable, trifunctionally bonded amide phase that is stable from pH 2 to 12 to enable the separation of polar analytes that span a wide range of polarity, structural moiety and pKa.
Catalog No. 50-466-116