Learn More
Waters Corp Column, Amide, 1.7 μm Particle Size, 2.1 mm x 50 mm
Supplier:  Waters Corp 176001907
Encompass_Preferred
ACQUITY UPLC BEH Amide columns provide stable HILIC separation of polar compounds. Utilizing BEH particle technology and a chemically stable amide phase (pH 2-12), they ensure efficient, repeatable separations with enhanced MS response.
  • Retains extremely polar compounds (e.g., carbohydrates, sugars).
  • Chemically stable amide phase (pH 2-12); ensures efficient UPLC HILIC separations.
  • Manufactured in cGMP, ISO 9001 accredited facility with rigorous batch testing.
  • Enhanced MS response and direct compatibility with sample preparation eluates.
  • Orthogonal selectivity; features eCord™ intelligent chip.
  • High resolution, high speed carbohydrate analysis via 1.7 μm particle size.
  • Increased chemical stability permits high pH/temperature use for faster analysis.
  • Exceptional column lifetime, improved robustness, prevents Schiff base formation.
Catalog No. 50-239-1596