Learn More
Waters Corp ACQUITY UPLC BEH C18 Column, 130Å, 1.7 µm, 2.1 mm X 30 mm, 1/pk
Supplier:  Waters Corp 186002349
Encompass_Preferred
As the premier column for method development, the 1.7 um ACQUITY UPLC BEH C18 sorbent provides you with the most universal column choice for UPLC separations. The trifunctionally bonded BEH particle gives you the widest usable pH range (pH 1-12), superior low pH stability, and ultra-low column bleed for high sensitivity MS applications.
Catalog No. 50-781-083