Learn More
Waters Corp Waters Corporation UPLC Column, BEH C18, 1.7 μm, 3 mm x 50 mm
Supplier:  Waters Corp 186004660
Encompass_Preferred
The ACQUITY UPLC BEH C18 Column, with its 1.7 μm BEH C18 sorbent, is ideal for UPLC/UHPLC separations. Its trifunctionally bonded BEH particle provides a wide pH range (1-12), superior low pH stability, and ultra-low column bleed for high sensitivity MS. It delivers high efficiency, peak symmetry, and chemical stability.
  • Designed for UPLC and UHPLC systems.
  • BEH Technology offers enhanced efficiency, strength, and pH range (1-12).
  • Superior low pH stability and ultra-low column bleed for MS applications.
  • Produced in cGMP, ISO 9001 certified plant using ultra-pure reagents.
  • Individually tested; Performance Chromatogram and Certificate of Batch Analysis provided via eCord.
  • Packing material batches chromatographically tested for reproducible performance.
Catalog No. 50-466-024