Learn More
Waters Corp Waters Corporation Column, C8, 130Å, 1.7 μm, 2.1 mm X 150 mm
Supplier:  Waters Corp 186003377
Encompass_Preferred
The ACQUITY UPLC BEH C8 column utilizes BEH C8 sorbent, which is less retentive than BEH C18, improving peak shape and chromatographic performance. Its trifunctionally bonded BEH particles ensure a wide usable pH range of 1–12, superior low pH stability, and ultra-low column bleed. This column delivers exceptional peak symmetry, efficiency, and chemical stability for diverse analytes.
  • Less retentive than C18 sorbent for faster elution and alternate selectivity.
  • Trifunctionally bonded BEH particles for wide usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • Exceptional peak symmetry, efficiency, and chemical stability.
  • Withstands pressures of up to 18,000 psi (1241 bar).
  • Designed for ultra-low dispersion UPLC systems.
  • eCord enabled.
Catalog No. 50-781-726