Learn More
Waters Corp Waters ACQUITY UPLC Column, C8, 1.7 μm, 2.1 mm × 100 mm
Supplier:  Waters Corp 186002878
Encompass_Preferred
This ACQUITY UPLC BEH C8 Column is an advanced analytical chromatography column designed for separating complex mixtures, particularly moderately non-polar compounds. It features C8 chemistry with an octadecyl stationary phase and operates using reversed-phase chromatography. Its trifunctionally bonded BEH particles offer a wide usable pH range (1 to 12 pH), superior low pH stability, and ultra-low column bleed, making it suitable for diverse UPLC applications.
  • C8 chemistry for reversed phase separation
  • Hybrid BEH particle technology with wide pH range (1 to 12 pH)
  • Withstands maximum pressure of 18000 psi (1240 Bar)
  • 1.7 μm particle size and 130 Å pore size for high resolution
  • Enhanced column stability and peak shape
  • Compatible with UPLC and UHPLC systems
Catalog No. 50-781-419