Learn More
Waters Corp WATERS ACQUITY UPLC BEH HILIC Column, 1.7 μm, 3 mm
Supplier:  Waters Corp 176001812
Encompass_Preferred
The ACQUITY UPLC BEH HILIC Column is designed for the retention and separation of very polar compounds, featuring an unbonded BEH substrate. These columns are optimized and tested to provide efficient, reproducible separations under HPLC HILIC conditions. They offer enhanced ruggedness compared to silica-based HILIC phases, with the BEH particle's chemical stability contributing to long column lifetimes.
  • Designed to retain and separate very polar compounds.
  • Optimized and tested for efficient, reproducible separations under HPLC HILIC conditions.
  • More rugged than silica-based HILIC phases.
  • BEH particle's chemical stability ensures long column lifetimes.
  • Includes eCord for enhanced functionality.
Catalog No. 50-791-045