Learn More
Waters Corp ACQUITY UPLC BEH HILIC Column, 130Å, 1.7 μm, 1 mm x 150 mm, 3 Pack
Supplier:  Waters Corp 176001091
Encompass_Preferred
ACQUITY UPLC BEH HILIC Columns enhance retention of polar species and provide an orthogonal separation for polar and ionizable compounds. These columns offer superior chemical stability and peak shape compared to silica-based HILIC stationary phases, using an acetonitrile-rich, low-aqueous mobile phase.
  • Enhanced retention of polar basic analytes.
  • Orthogonal separation for polar and ionizable compounds.
  • Exceptional chemical stability and peak shape versus silica-based HILIC.
  • Utilizes acetonitrile-rich, low-aqueous mobile phase.
  • Analytes elute in order of increasing hydrophilicity.
  • Improved separation and characterization of polar lipids for LC/MS studies.
Catalog No. 50-780-029