Learn More
Waters Corp ACQUITY UPLC BEH HILIC Column, 130Å, 1.7 µm, 2.1 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186003462
Encompass_Preferred
ACQUITY BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions. More rugged than silica based HILIC phases, the BEH particle’s chemical stability results in long column lifetimes.
Catalog No. 50-781-794