Learn More
Waters Corp Waters ACQUITY UPLC BEH HILIC Column, 130Å, 1.7 μm, 3 mm x 100 mm
Supplier:  Waters Corp 186004676
Encompass_Preferred
ACQUITY BEH HILIC columns, with unbonded BEH substrate, are designed for separating very polar compounds. Optimized for efficient, reproducible HPLC HILIC separations, they are more rugged than silica-based phases, ensuring long column lifetimes. They provide excellent pH stability (1–9) and withstand pressures up to 18,000 psi (1240 Bar).
  • Separates very polar compounds.
  • Efficient, reproducible HPLC HILIC separations.
  • Rugged, long-lasting compared to silica-based phases.
  • Excellent pH stability (1–9).
  • Withstands up to 18,000 psi (1240 Bar).
Catalog No. 50-791-185