Learn More
Waters Corp Waters Column, Phenyl, 1.7 μm, 2.1 mm, 100 mm
Supplier:  Waters Corp 186002885
Encompass_Preferred
ACQUITY BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This sorbent offers the most chemically stable phenyl sorbent on the market, providing ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Manufactured under strict ISO 9001 conditions and stringent process control using the trifunctionally-bonded phenyl-hexyl ligand.
  • Offers industry-leading pH stability, batch-to-batch reproducibility, and exceptional peak shape for all analyte types.
  • Provides complementary selectivity to straight-chain-alkyl columns, particularly for analytes containing aromatic rings.
  • Ensures ultra-low column bleed and long column lifetime.
  • Suitable for analysis of analytes with a pore size of 130Å and a particle size of 1.7 μm, and provides a wide pH stability range (1 to 12).
Catalog No. 50-781-425