Learn More
Waters Corp Waters Column, BEH Phenyl, 1.7 μm, 3 mm × 100 mm
Supplier:  Waters Corp 186004673
Encompass_Preferred
ACQUITY BEH Phenyl columns offer complementary selectivity to C18 stationary phases, ideal for polyaromatic compounds. They feature a trifunctionally bound phenyl-hexyl ligand, providing exceptional chemical stability, reproducibility, and excellent peak shape with ultra-low column bleed and long lifetimes.
  • Provides complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.
  • Offers the most chemically stable phenyl sorbent on the market.
  • Provides ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Industry-leading batch-to-batch reproducibility for all analyte classes.
  • Packing materials manufactured under exacting ISO 9001 standards, ensuring quality.
  • Each column is independently examined on the eCord™ intelligent chip.
Catalog No. 50-791-182