Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters SEC Column, 125 Å, 1.7 μm, 4.6 mm X 300 mm

Supplier:  Waters Corp 186006506

Encompass_Preferred

This column analyzes proteins and their aggregates, offering up to 10 times faster and more accurate aggregate determinations than traditional HPLC-based SEC methods. It provides improved component resolution, reduced analysis time, and lower mobile phase consumption in UPLC SEC applications. Utilizing sub-2-μm BEH particle technology, it is optimized for molecular weights from 1,000 to 80,000 daltons.

  • Chemistry: SEC
  • Separation mode: SEC
  • Particle substrate: Hybrid
  • pH range: 2.5 to 8 pH
  • Temperature limits: 60°C
  • Max pressure: 7000 psi (483 bar)
  • MW range: 1000 to 80000 daltons
  • Particle size: 1.7 μm
  • Pore size: 125 Å
  • Format: Column
  • System: UPLC, UHPLC
  • Application: Protein
  • Technology: Unique Ethylene-Bridged Hybrid (BEH) Diol coated particle technology.

Catalog No. 50-201-650


May include imposed supplier surcharges.
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.