Learn More
Waters Corp ACQUITY UPLC Peptide BEH C18 Column, 300Å Pore Size, 1.7 μm Particle Size
Supplier:  Waters Corp 186003685
Encompass_Preferred
The ACQUITY UPLC Peptide BEH C18 Columns are specifically designed and quality control tested for peptide separations, making them an ideal choice for proteomics, protein characterization, and peptide synthesis. These columns utilize a wide-pore (300Å) trifunctionally bonded BEH particle, which provides an extensive usable pH range, excellent stability at low pH, and minimal column bleed.
  • Optimized and QC tested for peptide separations.
  • Wide-pore (300Å) trifunctionally bonded BEH particle.
  • Ultra-low column bleed.
  • Produces narrow, symmetrical peaks for best resolution.
  • Achieves successful separations of a wide range of peptides.
  • Unique column chemistry combined with sub-2 μm particles ensures ultimate resolution.
  • Particle structure and bonding chemistry provide pH stability from 1 to 12.
Catalog No. 50-781-951