Learn More
Waters Corp ACQUITY UPLC Peptide BEH C18 Column, 300Å, 1.7 µm, 2.1 mm X 100 mm, 1/pk
Supplier:  Waters Corp 186003686
Encompass_Preferred
The ACQUITY BEH300 C18 Peptide Separation Technology columns have been specially optimized and QC tested for peptide separations. The wide-pore (300Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
Catalog No. 50-781-952