Learn More
Waters Corp Waters ACQUITY UPLC Peptide BEH C18 Method Validation Kit, 300Å, 1.7 μm, 2.1 mm x 150 mm, 3 pk
Supplier: Waters Corp 186006517
This method validation kit includes three columns packed with three different batches of sorbent, designed for reliable performance in UPLC and UHPLC systems.
- C18 chemistry provides reversed phase separation.
- Hybrid particle substrate.
- Operates within a pH range of 1 to 12.
- Endcapped for enhanced performance.
- Low silanol activity.
- Suitable for molecular weights ranging from 1000 to 30,000.
- Spherical particle shape with a size of 1.7 μm.
- Parker-style endfitting.
- Pore size of 300 Å.
- Quality control tested for peptide applications.
- Surface area of 185.
- Utilizes BEH particle technology.
- USP classification L1.
- Inner diameter of 2.1 mm and length of 150 mm.
- Carbon load of 18%.
- Package contains 3 units.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.