Learn More
Waters Corp Waters ACQUITY UPLC Protein BEH C4 Column, 300 Å, 1.7 μm, 1 mm x 150 mm
Supplier:  Waters Corp 186005591
Encompass_Preferred
The ACQUITY UPLC Protein BEH C4, 300 Å, 1.7 μm, 1 mm x 150 mm column is QC tested and optimized to separate proteins based on size, hydrophobicity and isoelectric point. The C4 ligand is less retentive compared to traditional C18 phases, which helps to minimize protein carryover, increase protein recovery, and improve peak capacity.
  • Optimized for separating proteins based on size, hydrophobicity, and isoelectric point.
  • C4 ligand minimizes protein carryover, increases protein recovery, and improves peak capacity.
  • Hybrid particle substrate with C4 chemistry, 300 Å pore size, and 1.7 μm particle size.
  • Dimensions: 1 mm inner diameter x 150 mm length.
  • Supports reversed phase separation mode, usable within a pH range of 2 to 10.
Catalog No. 50-466-134