Learn More
Waters Corp ACQUITY UPLC Protein BEH SEC Column, 450Å, 2.5 µm, 4.6 mm X 150 mm, 100K - 1500K, 1/pk
Supplier:  Waters Corp 186006851
Encompass_Preferred
Based upon the highly successful BEH particle technology, the ACQUITY UPLC BEH450 SEC 2.5 µm column is fully optimized for the analysis of proteins and their aggregates with molecular weights ranging from 100,000 to 1,500,000 Daltons, the total system solution will routinely deliver fast, accurate aggregate determinations up to 10 times quicker that traditional HPLC-based SEC methods.
Catalog No. 50-466-511