Learn More
Waters Corp ACQUITY UPLC Column, Peptide CSH C18 1.7 μm, 2.1 mm x 150 mm
Supplier:  Waters Corp 186006938
Encompass_Preferred
The ACQUITY UPLC Peptide CSH C18 column features 1.7 μm particles with a low and carefully-defined concentration of positive charges, which yields excellent peak shape for peptides in FA- or TFA-containing mobile phases. Its design makes it highly suitable for LC or LC-MS applications.
  • Low, carefully-defined positive charges optimize peak shape for peptides in FA- or TFA-containing mobile phases.
  • Compatible with standard TFA-containing eluents or weaker acid modifiers.
  • Accepts greater peptide mass loads, improving detection of low-level constituents.
  • CSH Technology delivers improved sample mass loadability and superior performance.
  • Excellent for challenging separations of mixtures with vastly different concentrations.
  • Supports high peak capacity peptide separations using UPLC and HPLC.
  • QC tested for column reproducibility.
Catalog No. 50-596-894