Learn More
Waters Corp ACQUITY UPLC CSH C18 Column, 130Å, 1.7 µm, 2.1 mm X 75 mm, 1/pk
Supplier:  Waters Corp 186005620
Encompass_Preferred
ACQUITY CSH C18 columns were designed to provide alternate selectivity compared to other reversed-phase UPLC columns. These ultra-efficient columns overcome peak shape asymmetry and poor loading capacity for basic compounds, column bleed, and rapid mobile-phase re-equilibration.
Catalog No. 50-466-156