Learn More
Waters Corp UPLC CSH C18 Column, 1.7 μm, 130 Å, 3 mm × 100 mm
Supplier:  Waters Corp 186005301
Encompass_Preferred
ACQUITY CSH C18 columns offer alternative selectivity for reversed-phase UPLC. Leveraging Charged Surface Hybrid (CSH) Technology, they resolve issues like peak shape asymmetry, poor loading capacity for basic compounds, column bleed, and slow mobile-phase re-equilibration, thereby enhancing chromatographic performance.
  • Resolves peak shape asymmetry and improves loading for basic compounds.
  • Minimizes column bleed and accelerates mobile-phase re-equilibration.
  • Ensures good peak shape and seamless transferability across formats.
  • Achieves TFA-like peak shape performance without Trifluoroacetic acid.
  • Simplifies method development using MS-friendly mobile phase additives.
  • Increases analytical loadability for sharper basic compound peaks.
  • Suitable for isolation and purification, reducing solvent use.
Catalog No. NC0826780