Learn More
Waters Corp ACQUITY UPLC CSH Fluoro-Phenyl Column, 1.7 μm, 3 x 75 mm
Supplier:  Waters Corp 186005625
Encompass_Preferred
This Fluoro-Phenyl column is designed to maximize selectivity differences, providing maximum method development freedom, especially when using low pH mobile phases. Its non-encapped sorbent provides enhanced retention of acidic compounds.
  • Provides exceptional selectivity for positional isomers, halogenated compounds, and polar compounds.
  • Utilizes multiple retention mechanisms, including dipole-dipole, hydrogen-bonding, aromatic (π–π), and hydrophobic interactions.
  • Offers enhanced retention of acidic compounds due to unique Charged Surface Hybrid (CSH) particle chemistry.
  • Delivers unique column selectivity with industry-leading reproducibility.
  • Ensures exceptional peak shape and loading capacity for basic compounds at low and high pH without ion-pair reagents.
  • Provides exceptional stability and advanced column equilibration at low and high pH.
Catalog No. 50-466-161