Learn More
Waters Corp Waters Column, Fluoro-Phenyl, 1.7 μm, 2.1 mm × 50 mm
Supplier:  Waters Corp 186005351
Encompass_Preferred
This CSH Fluoro-Phenyl column provides maximum selectivity differences and method development freedom, especially with low pH mobile phases. Its non-encapped sorbent enhances acidic compound retention. Exceptional selectivity for isomers, halogenated, and polar compounds is achieved via multiple retention mechanisms. Based on third-generation hybrid particle technology, it offers improved sample loadability, peak asymmetry, mechanical, and chemical stability. An eCord Intelligent Chip tracks performance history.
  • Superior peak shape for basic compounds.
  • Increased loading capacity and rapid column equilibration.
  • Improved reproducibility.
  • Exceptional stability at both low and high pH.
  • Designed for use with UPLC systems.
  • Manufactured in cGMP, ISO 9001 certified facilities.
  • Each batch tested with acidic, basic, and neutral analytes, results held to narrow specifications.
Catalog No. 50-203-7701