Learn More
Waters Corp Waters ACQUITY UPLC CSH Phenyl-Hexyl Column, 1.7 μm, 3.0 x 75 mm
Supplier: Waters Corp 186005624
ACQUITY UPLC Columns are designed to integrate seamlessly with ACQUITY UPLC Systems. The CSH Phenyl-Hexyl Column features sub-2-μm, fully porous particles, providing high efficiencies and wide selectivity. Its ethylene-bridged hybrid particles offer superior peak shape for basic analytes without ion-pairing reagents, ensuring robust performance and ultimate flexibility for method development.
- Sub-2-μm fully porous particle technology.
- High efficiencies and wide selectivity.
- Best-in-class column stability.
- Optimal for high-throughput method development.
- Provides superior peak shape for basic analytes with acidic mobile phases.
- Eliminates need for ion-pairing reagents.
- Particle size: 1.7 μm.
- Dimensions: 3.0 x 75 mm.
- pH range: 1 to 11.
- Temperature limits: 80°C (low pH), 45°C (high pH).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.