Learn More
Waters Corp Glycoprotein BEH HILIC Column, 300Å, 1.7 μm, 2.1 mm x 5 mm
Supplier: Waters Corp 186007960
These columns are primarily designed for the analysis of intact glycoproteins, glycoprotein fragments, and glycopeptides. Each batch of Glycoprotein BEH Amide 300Å, 1.7μm material is specifically quality control tested with a Glycoprotein Performance Test Standard to help ensure batch to batch consistency.
- HILIC chemistry with hydrophilic interaction separation.
- Hybrid particle substrate with spherical shape.
- pH range of 2 to 10.
- Particle size: 1.7 μm.
- Direct-connect endfitting.
- Pore size: 300 Å.
- Glycoprotein QC tested for UHPLC and UPLC systems.
- BEH particle technology.
- Dimensions: 2.1 mm inner diameter, 5 mm length.
- Carbon load: 12%.
- Glycan application.
- Package contains 3 units.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.