Learn More
Waters Corp Waters ACQUITY UPLC HSS C18 Column, 100Å, 1.8 μm, 2.1 mm X 150 mm
Supplier:  Waters Corp 186003534
Encompass_Preferred
ACQUITY UPLC HSS C18 chemistry provides an ultra-performance general purpose C18 bonded phase, delivering superior peak shape for bases, increased retention, and long column lifetimes at low pH. These columns are fully scalable and transferable to HPLC separations.
  • High Strength Silica (HSS) particle technology offers mechanical stability for UPLC pressures and increased retention.
  • Trifunctionally bonded C18 ligand and proprietary end-capping prevent acid hydrolysis, ensuring excellent performance under acidic conditions.
  • Fully end-capped design provides superior peak shape for bases and enhanced retention.
  • Manufactured in a cGMP, ISO 9001:2000 certified plant, ensuring quality and consistent specifications.
  • HSS technology ensures high efficiency, sharp peaks, faster analyses, and minimizes analyte adsorption for greater sensitivity.
Catalog No. 50-781-824