Learn More
Waters Corp ACQUITY UPLC HSS C18 Method Validation Kit, 100Å, 1.8 µm, 2.1 mm X 50 mm, 3/pk
Supplier:  Waters Corp 186004057
Encompass_Preferred
This kit consists of three ACQUITY HSS C18 columns packed with three different batches of sorbent.
Catalog No. 50-782-162